
n-Octyl β-D-maltoside

Synonym(s):
n-Octyl β-D-maltopyranoside
Empirical Formula (Hill Notation):
C20H38O11
CAS Number:
Molecular Weight:
454.51
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing