
N-Benzoyl-L-tyrosine p-nitroanilide

Empirical Formula (Hill Notation):
C22H19N3O5
CAS Number:
Molecular Weight:
405.40
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing