
Milrinone

Synonym(s):
1,6-Dihydro-2-methyl-6-oxo-(3,4′-bipyridine)-5-carbonitrile
Empirical Formula (Hill Notation):
C12H9N3O
CAS Number:
Molecular Weight:
211.22
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade