
Methimazole Related Compound A

Synonym(s):
(Methylamino)acetaldehyde dimethyl acetal, 1,1-Dimethoxy-2-methylaminoethane, 2,2-Dimethoxy-N-methylethylamine
Linear Formula:
CH3NHCH2CH(OCH3)2
CAS Number:
Molecular Weight:
119.16
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2