Skip to Content
Merck
Maprotiline Hydrochloride pharmaceutical secondary standard, certified reference material

Maprotiline hydrochloride

Maprotiline Hydrochloride pharmaceutical secondary standard, certified reference material
Synonym(s):
9-(γ-Methylaminopropyl)-9,10-dihydro-9,10-ethanoanthracene hydrochloride, Maprotiline hydrochloride
Empirical Formula (Hill Notation):
C20H23N · HCl
CAS Number:
Molecular Weight:
313.86

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade