Skip to Content
BF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lectin from
Triticum vulgaris
(wheat)
Synonym(s):
WGA, Wheat germ agglutinin
Products
Brand
Sigma-Aldrich (7)
Physical Form
Chemical Composition
Conjugate
Quality Segment
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
L4895
Lectin from
Triticum vulgaris
(wheat),
FITC conjugate, lyophilized powder
Expand
View Pricing
L9640
Lectin from
Triticum vulgaris
(wheat),
lyophilized powder
Expand
View Pricing
L5142
Lectin from
Triticum vulgaris
(wheat),
biotin conjugate, lyophilized powder
Expand
View Pricing
L3892
Lectin from
Triticum vulgaris
(wheat),
peroxidase conjugate, lyophilized powder
Expand
View Pricing
L0636
Lectin from
Triticum vulgaris
(wheat),
suitable for cell culture, BioReagent, lyophilized powder
Expand
View Pricing
L1882
Lectin from
Triticum vulgaris
(wheat),
agarose conjugate, saline suspension
Expand
View Pricing
L1394
Lectin from
Triticum vulgaris
(wheat),
agarose conjugate, saline suspension
Expand
View Pricing