
Isopropenylmagnesium bromide solution

Linear Formula:
CH2=C(CH3)MgBr
CAS Number:
Molecular Weight:
145.28
Products
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing