
Hemocyanin from Megathura crenulata (keyhole limpet)

Synonym(s):
KLH, Keyhole limpet hemocyanin
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Quality Segment
Package Size
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing