
Guanfacine hydrochloride

Synonym(s):
N (Aminoiminomethyl)-2,6-dichlorobenzeneacetamide, Guanfacine hydrochloride
Empirical Formula (Hill Notation):
C9H9Cl2N3O · HCl
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique