
Ginsenoside Rg5

Synonym(s):
(3β, 12β,20E)-12-Hydroxydammara-20(22),24-dien-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C42H70O12
CAS Number:
Molecular Weight:
767.00
Products
Brand
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing