Skip to Content
BF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
GC Stationary Phase
Products
Brand
Supelco (10)
Physical Form
Agency
Column Type
Quality Segment
Package Size
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
21032
GC Stationary Phase,
phase Carbowax 20M, bottle of 50 g
Expand
View Pricing
21228
GC Stationary Phase,
phase OV-101, bottle of 20 g
Expand
View Pricing
21104
GC Stationary Phase,
phase OV-1, bottle of 10 g
Expand
View Pricing
21123
GC Stationary Phase,
phase Triton X-100, bottle of 50 g
Expand
View Pricing
21284-U
GC Stationary Phase,
phase SP
®
-2100, bottle of 10 g
Expand
View Pricing
21106
GC Stationary Phase,
phase SE-54, bottle of 50 g
Expand
View Pricing
21028
GC Stationary Phase,
phase Carbowax 1540, pkg of 50 g
Expand
View Pricing
21217
GC Stationary Phase,
phase 1,2,3-tris(2-Cyanoethoxy)propane (TCEP), bottle of 50 g
Expand
View Pricing
21098-U
GC Stationary Phase,
phase DC QF-1 (FS 1265), bottle of 50 g
Expand
View Pricing
85427
GC Stationary Phase,
phase DC-710, bottle of 100 mL
Expand
View Pricing