
Ethylenedinitrilotetraacetic acid disodium salt dihydrate

Synonym(s):
Ethylenediaminetetraacetic acid disodium salt dihydrate, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2
Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Reaction Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing