
Ethylenediaminetetraacetic acid iron(III) sodium salt

Synonym(s):
EDTA iron(III) sodium salt, Edathamil, Komplexon™ II
Linear Formula:
C10H12N2NaFeO8
Products
Brand
Biological Source
Formula Weight
Physical Form
Agency
Chemical Composition
Quality Segment
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing