Skip to Content
BF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Estapor
®
Magnetic Microspheres
Products
Brand
Sigma-Aldrich (5)
Color
Chemical Composition
Quality Segment
Functional Group
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
FR123710087
Estapor
®
Magnetic Microspheres,
EM1-100/40 Mag COOH PS 10%
, Encapsulated Magnetic carboxylated microspheres
1µm,-Available in 0.5g, 1g, 10g, 100g - For ordering, click "Request more information"
Expand
View Pricing
FR180380000
Estapor
®
Magnetic Microspheres,
M1-030/40 Mag COOH PS 10%,Magnetic microspheres
300nm
, ChemiLuminsecence Immunoassay, Lateral Flow Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR139572001
Estapor
®
Magnetic Microspheres,
EM2-100/40 Mag
NH2
PS 10%, Encapsulated Magnetic aminated microspheres
1µm
, ChemiLuminsecence Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR123709083
Estapor
®
Magnetic Microspheres,
M1-070/40 Mag
COOH
PS 10%, Magnetic carboxylated microspheres
1µm
, ChemiLuminsecence Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR180380073
Estapor
®
Magnetic Microspheres,
M1-020/50 Mag
COOH
PS 10%, Magnetic microspheres
200nm
, ChemiLuminsecence Immunoassay, Lateral Flow Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR180380074
Estapor
®
Magnetic Microspheres,
MS-070/40 Mag PS 10%, Magnetic microspheres
1µm,
ChemiLuminsecence Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR123711081
Estapor
®
Magnetic Microspheres,
M1-070/60 Mag
COOH
PS 10%, Magnetic carboxylated microspheres
1µm
, ChemiLuminsecence Immunoassay - For ordering, click "Request more information"
Expand
View Pricing