
Eppendorf® twin.tec PCR Plate 96 LoBind

Synonym(s):
Eppendorf® twin.tec® PCR Plate 96 LoBind, Eppendorf twin.tec® PCR Plates 96
Products
Product Category
Color
Manufacturer
Binding Type
Color
Sterility
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing