
EPA 547 Glyphosate Solution

Synonym(s):
Glyphosate solution, N-(Phosphonomethyl)glycine
Products
Product Category
Brand
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing