Skip to Content
Merck
Edetate Calcium Disodium Pharmaceutical Secondary Standard; Certified Reference Material

Edetate Calcium Disodium

Edetate Calcium Disodium Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Ethylenediaminetetraacetic acid calcium disodium salt hydrate, Calcium disodium ethylenediaminetetraacetate hydrate
Linear Formula:
Ca(OOCCH2)2NCH2CH2N(CH2COONa)2 · aq
CAS Number:
Molecular Weight:
374.27 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique