
Ecamsule Related Compound A

Synonym(s):
1,4-Benzenedimethanol, p-Phenylene dicarbinol, p-Phenylenedimethanol, p-Xylene-α,α′-diol, p-Xylylene dialcohol, NSC 5097
Linear Formula:
C6H4(CH2OH)2
CAS Number:
Molecular Weight:
138.16
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing