
Doxorubicin hydrochloride

Synonym(s):
DOX, Hydroxydaunorubicin hydrochloride, Doxorubicin hydrochloride
Empirical Formula (Hill Notation):
C27H29NO11 · HCl
CAS Number:
Molecular Weight:
579.98
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing