
(±)-Enterolactone

Synonym(s):
Enterolactone, trans-α,β-Bis(3-hydroxybenzyl)butyrolactone
Empirical Formula (Hill Notation):
C18H18O4
CAS Number:
Molecular Weight:
298.33
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing