Skip to Content
Merck
Digitoxin British Pharmacopoeia (BP) Reference Standard

Digitoxin

Digitoxin British Pharmacopoeia (BP) Reference Standard
Synonym(s):
5β,20(22)-Cardenolide-3β,14-diol-3-(O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy, Digitoxoside, Lanatoxin
Empirical Formula (Hill Notation):
C41H64O13
CAS Number:
Molecular Weight:
764.94
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade