Dichloro(p-cymene)ruthenium(II) dimer
Synonym(s):
(p-Cymene)ruthenium(II) chloride dimer, Benzene, 1-methyl-4-(1-methylethyl)-, ruthenium complex
Linear Formula:
[Ru(p-cymene)Cl2]2
CAS Number:
Molecular Weight:
612.39
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing