
Desogestrel Related Compound B

Synonym(s):
3-Hydroxy-desogestrel, (3β,17α)-13-Ethyl-11-methylene-18,19-dinorpregn-4-en-20-yne-3,17-diol
Empirical Formula (Hill Notation):
C22H30O2
CAS Number:
Molecular Weight:
326.47
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing