Cyromazin
Synonym(s):
N-Cyclopropyl-1,3,5-triazin-2,4,6-triamine, Cyromazin
Empirical Formula (Hill Notation):
C6H10N6
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2