
Corticotropin Releasing Factor sheep

Synonym(s):
CRF, CRH
Empirical Formula (Hill Notation):
C205H339N59O63S
CAS Number:
Molecular Weight:
4670.31
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing