
Clarithromycin Impurity R

Synonym(s):
DEMA, N-Demethylerythromycin A
Empirical Formula (Hill Notation):
C36H65NO13
CAS Number:
Molecular Weight:
719.90
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing