
Cimetidine

Synonym(s):
SKF-92334
Empirical Formula (Hill Notation):
C10H16N6S
CAS Number:
Molecular Weight:
252.34
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing