
Ciclopirox Related Compound B

Synonym(s):
6-Cyclohexyl-4-methyl-2-pyrone
Empirical Formula (Hill Notation):
C12H16O2
CAS Number:
Molecular Weight:
192.25
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2