Skip to Content
BF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chloroacetic acid
Synonym(s):
Monochloroacetic acid, Chloroacetic acid
Linear Formula:
ClCH
2
COOH
CAS Number:
79-11-8
Molecular Weight:
94.50
EC Number:
201-178-4
Products
Product Category
organic acids (2)
pesticides (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
ACS reagent (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
402923
Chloroacetic acid,
ACS reagent, ≥99.0%
Expand
View Pricing
8.00412
Chloroacetic acid,
for synthesis
Expand
View Pricing
C19627
Chloroacetic acid,
99%
Expand
View Pricing
36544
Chloroacetic acid,
PESTANAL
®
, analytical standard
Expand
View Pricing