![Cetirizine Related Compound G certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/186/411/fa1259ab-f206-468a-9643-c3a9d867ae8d/640/fa1259ab-f206-468a-9643-c3a9d867ae8d.jpg)
Cetirizine Related Compound G
![Cetirizine Related Compound G certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/186/411/fa1259ab-f206-468a-9643-c3a9d867ae8d/640/fa1259ab-f206-468a-9643-c3a9d867ae8d.jpg)
Synonym(s):
2-{4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl}ethanol dihydrochloride
Empirical Formula (Hill Notation):
C19H23ClN2O · 2HCl
CAS Number:
Molecular Weight:
403.77
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2