Skip to Content
Merck
Carbon nanofibers pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm

Carbon nanofibers

Carbon nanofibers pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Synonym(s):
Conical carbon nanofibers
Empirical Formula (Hill Notation):
C
Molecular Weight:
12.01

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Tissue
Quality Segment
Pore Size (Å)