
Arachidyl alcohol

Synonym(s):
1-Eicosanol, Arachinyl alcohol
Linear Formula:
CH3(CH2)19OH
CAS Number:
Molecular Weight:
298.55
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing