
Apocynin

Synonym(s):
Apocynin, 4′-Hydroxy-3′-methoxyacetophenone, Acetovanillone
Linear Formula:
HOC6H3(OCH3)COCH3
CAS Number:
Molecular Weight:
166.17
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Research Area
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing