
Almotriptan malate

Synonym(s):
3-[2-(Dimethylamino)ethyl]-5-(pyrrolidin-1-ylsulfonylmethyl )-1H-indole malate, PNU 180638
Empirical Formula (Hill Notation):
C17H25N3O2S · C4H6O5
CAS Number:
Molecular Weight:
469.55
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3