
Alizarin

Synonym(s):
1,2-Dihydroxyanthraquinone, Mordant Red 11
Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing