Skip to Content
Merck
Adenosine 5′-triphosphate (ATP) disodium salt hydrate vial of ~1 mg ATP

Adenosine 5′-triphosphate (ATP) disodium salt hydrate

Adenosine 5′-triphosphate (ATP) disodium salt hydrate vial of ~1 mg ATP
Synonym(s):
ATP disodium hydrate
Linear Formula:
C10H14N5O13P3Na2 · xH2O
CAS Number:
Molecular Weight:
551.14 (anhydrous basis)
EC Number:

Products

Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Package Size