Skip to Content
Merck
α-Damascone analytical standard

α-Damascone

α-Damascone analytical standard
Synonym(s):
1-(2,6,6-Trimethyl-2-cyclohexen-1-yl)-2-buten-1-one, 2,6,6-Trimethyl-1-crotonyl-2-cyclohexene
Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30

Products

Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing