
Acetaldehyde-2,4-dinitrophenylhydrazone

Linear Formula:
CH3CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
224.17
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2