
8arm-PEG15K

Synonym(s):
8arm-PEG-Succinimidyl Glutarate
Linear Formula:
R(O(CH2CH2O)nCOCH2CH2CH2COOC4H4O2N)8
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing