
6a-Hydroxyhydrocortisone

Synonym(s):
6α-Hydroxycortisol, 4-Pregnene-6α,11β,17,21-tetrol-3,20-dione
Empirical Formula (Hill Notation):
C21H30O6
CAS Number:
Molecular Weight:
378.46
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing