
5-Amino-3-methyl-isothiazole hydrochloride

Empirical Formula (Hill Notation):
C4H6N2S · HCl
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing