Skip to Content
Merck
3,5-Bis(trifluoromethyl)benzyl bromide 97%

3,5-Bis(trifluoromethyl)benzyl bromide

3,5-Bis(trifluoromethyl)benzyl bromide 97%
Synonym(s):
1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene
Linear Formula:
(CF3)2C6H3CH2Br
CAS Number:
Molecular Weight:
307.03
EC Number:

Products

Product Category
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing