![3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate 98%](/deepweb/assets/sigmaaldrich/product/structures/322/134/88b00489-fbbd-47b7-ba3e-430dd6a3c852/640/88b00489-fbbd-47b7-ba3e-430dd6a3c852.png)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate
![3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate 98%](/deepweb/assets/sigmaaldrich/product/structures/322/134/88b00489-fbbd-47b7-ba3e-430dd6a3c852/640/88b00489-fbbd-47b7-ba3e-430dd6a3c852.png)
Synonym(s):
CHAPS
Empirical Formula (Hill Notation):
C32H58N2O7S · xH2O
CAS Number:
Molecular Weight:
614.88 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing