
2-Nitro-5-thiocyanatobenzoic acid

Synonym(s):
2-Nitro-5-thiocyanobenzoic acid, NTCB
Linear Formula:
O2NC6H3(SCN)CO2H
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing