Skip to Content
Merck
2-Keto-D-glucose ≥98.0% (TLC)

2-Keto-D-glucose

2-Keto-D-glucose ≥98.0% (TLC)
Synonym(s):
D-Glucosone
Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing