
2-(2-Ethylhexyloxy)ethanol

Synonym(s):
Ethylene glycol 2-ethylhexyl ether
Linear Formula:
CH3(CH2)3CH(C2H5)CH2OCH2CH2OH
CAS Number:
Molecular Weight:
174.28
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing