Skip to Content
BF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,2-Dimethoxypropane
Synonym(s):
Acetone dimethyl acetal, 2,2-Dimethoxypropane
Linear Formula:
(CH
3
)
2
C(OCH
3
)
2
CAS Number:
77-76-9
Molecular Weight:
104.15
EC Number:
201-056-0
Products
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
D136808
2,2-Dimethoxypropane,
reagent grade, 98%
Expand
View Pricing
8.02936
2,2-Dimethoxypropane,
for synthesis
Expand
View Pricing
73137
2,2-Dimethoxypropane,
analytical standard
Expand
View Pricing