
2,2′-Biquinoline-4,4′-dicarboxylic acid dipotassium salt trihydrate

Synonym(s):
2,2′-Bicinchoninic acid dipotassium salt, 4,4′-Dicarboxy-2,2′-biquinoline dipotassium salt, Dipotassium 2,2′-biquinoline-4,4′-dicarboxylate trihydrate
Empirical Formula (Hill Notation):
C20H10K2N2O4 · 3H2O
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing