Skip to Content
Merck
1-Piperidinepropionic acid 96%

1-Piperidinepropionic acid

1-Piperidinepropionic acid 96%
Synonym(s):
3-Piperidin-1-ylpropionic acid
Empirical Formula (Hill Notation):
C8H15NO2
CAS Number:
Molecular Weight:
157.21
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing