Skip to Content
Merck
1-Palmitoyl-sn-glycero-3-phosphocholine synthetic, ≥99%

1-Palmitoyl-sn-glycero-3-phosphocholine

1-Palmitoyl-sn-glycero-3-phosphocholine synthetic, ≥99%
Synonym(s):
1-Hexadecanoyl-sn-glycero-3-phosphocholine, 3-sn-Lysophosphatidylcholine, 1-palmitoyl, L-α-Palmitoyl-lysophosphatidylcholine, L-γ-Palmitoyl-α-lysolecithin, Lysolecithin, palmitoyl
Empirical Formula (Hill Notation):
C24H50NO7P
CAS Number:
Molecular Weight:
495.63

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing